Methyl-3-[(3-methoxypropyl)amino]propanoate

Supplier: Matrix Scientific

026781-500MG
101822-858EA 306.15 USD
101822-858
Methyl-3-[(3-methoxypropyl)amino]propanoate
Methyl-3-[(3-methoxypropyl)amino]propanoate

Hazard: Irritant

Formula: C₈H₁₇NO₃
MDL Number: MFCD10687247
CAS Number: 51935-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR