N1-(sec-Butyl)-1,3-propanediamine

Supplier: Matrix Scientific

026778-500MG
101822-852EA 306.15 USD
101822-852
N1-(sec-Butyl)-1,3-propanediamine
N1-(sec-Butyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₇H₁₈N₂
MDL Number: MFCD10687245
CAS Number: 854650-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR