3-(sec-Butylamino)propanenitrile

Supplier: Matrix Scientific

026777-500MG
101822-850EA 306.15 USD
101822-850
3-(sec-Butylamino)propanenitrile
3-(sec-Butylamino)propanenitrile

Hazard: Irritant

Formula: C₇H₁₄N₂
MDL Number: MFCD01475812
CAS Number: 21539-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR