3-(sec-Butylamino)propanamide

Supplier: Matrix Scientific

026776-500MG
101822-848EA 306.15 USD
101822-848
3-(sec-Butylamino)propanamide
3-(sec-Butylamino)propanamide

Hazard: Irritant

Formula: C₇H₁₆N₂O
MDL Number: MFCD10687244
CAS Number: 1040687-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR