3-[(3-Hydroxypropyl)amino]propanenitrile

Supplier: Matrix Scientific

026771-500MG
101822-838EA 306.15 USD
101822-838
3-[(3-Hydroxypropyl)amino]propanenitrile
3-[(3-Hydroxypropyl)amino]propanenitrile

Hazard: Irritant

Formula: C₆H₁₂N₂O
Boiling Pt: 110…115 °C
MDL Number: MFCD10687241
CAS Number: 34449-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR