3-(4-Morpholinyl)propanohydrazide

Supplier: Matrix Scientific

026768-500MG
101822-832EA 306.15 USD
101822-832
3-(4-Morpholinyl)propanohydrazide
3-(4-Morpholinyl)propanohydrazide

Hazard: Irritant

Formula: C₇H₁₅N₃O₂
Melting Pt: 66…69 °C
MDL Number: MFCD01838567
CAS Number: 59737-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR