2-Methyl-1-propyl-1H-indol-5-ylamine hydrochloride

Supplier: Matrix Scientific

026720-500MG
101822-718EA 495.38 USD
101822-718
2-Methyl-1-propyl-1H-indol-5-ylamine hydrochloride
2-Methyl-1-propyl-1H-indol-5-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD09997707
CAS Number: 1185297-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR