1-(2-Methoxyethyl)-2-methyl-1H-indol-5-ylamine hydrochloride

Supplier: Matrix Scientific

026719-500MG
101822-716EA 495.38 USD
101822-716
1-(2-Methoxyethyl)-2-methyl-1H-indol-5-ylamine hydrochloride
1-(2-Methoxyethyl)-2-methyl-1H-indol-5-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD09997706
CAS Number: 883545-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR