2-Azepan-1-yl-2-(4-chlorophenyl)ethylaminehemioxalate

Supplier: Matrix Scientific

026717-500MG
101822-712EA 306.15 USD
101822-712
2-Azepan-1-yl-2-(4-chlorophenyl)ethylaminehemioxalate
2-Azepan-1-yl-2-(4-chlorophenyl)ethylaminehemioxalate

Hazard: Irritant

Formula: C₃₀H₄₄Cl₂N₄O₄
MW: 595.62 g/mol
MDL Number: MFCD09997704
CAS Number: 1185089-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR