Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine hydrochloride

Supplier: Matrix Scientific

026714-500MG
101822-708EA 306.15 USD
101822-708
Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine hydrochloride
Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₃
MW: 245.71 g/mol
MDL Number: MFCD07110829
CAS Number: 1049803-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR