N'-Benzo[1,3]dioxol-5-ylmethyl-N,N-dimethylethane-1,2-diamine dihydrochloride

Supplier: Matrix Scientific

026713-500MG
101822-706EA 495.38 USD
101822-706
N'-Benzo[1,3]dioxol-5-ylmethyl-N,N-dimethylethane-1,2-diamine dihydrochloride
N'-Benzo[1,3]dioxol-5-ylmethyl-N,N-dimethylethane-1,2-diamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀Cl₂N₂O₂
MW: 295.21 g/mol
MDL Number: MFCD07110825
CAS Number: 128120-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR