C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine dihydrochloride

Supplier: Matrix Scientific

026709-500MG
101822-698EA 495.38 USD
101822-698
C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine dihydrochloride
C-(4-Chlorophenyl)-C-pyridin-3-ylmethylamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃Cl₃N₂
MDL Number: MFCD08544225
CAS Number: 883548-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR