4-(4-Methoxyphenoxymethyl)piperidine hydrochloride

Supplier: Matrix Scientific

026708-500MG
101822-696EA 306.15 USD
101822-696
4-(4-Methoxyphenoxymethyl)piperidine hydrochloride
4-(4-Methoxyphenoxymethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD09997701
CAS Number: 1135262-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR