2-Ethoxypropylamine hydrochloride

Supplier: Matrix Scientific

026707-500MG
101822-694EA 164.62 USD
101822-694
2-Ethoxypropylamine hydrochloride
2-Ethoxypropylamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₄ClNO
MDL Number: MFCD09997700
CAS Number: 1184979-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR