2-Methyl-3-(4-pyridin-2-ylpiperazin-1-yl)propionic acid dihydrochloride

Supplier: Matrix Scientific

026700-500MG
101822-682EA 306.15 USD
101822-682
2-Methyl-3-(4-pyridin-2-ylpiperazin-1-yl)propionic acid dihydrochloride
2-Methyl-3-(4-pyridin-2-ylpiperazin-1-yl)propionic acid dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁Cl₂N₃O₂
MDL Number: MFCD08541874
CAS Number: 1185304-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR