3-[4-(4-Methoxyphenyl)piperazin-1-yl]propionic acid dihydrochloride

Supplier: Matrix Scientific

026699-500MG
101822-680EA 306.15 USD
101822-680
3-[4-(4-Methoxyphenyl)piperazin-1-yl]propionic acid dihydrochloride
3-[4-(4-Methoxyphenyl)piperazin-1-yl]propionic acid dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂O₃
MDL Number: MFCD09997696
CAS Number: 1185294-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR