3-[4-(3-Chlorophenyl)piperazin-1-yl]propionic acid dihydrochloride

Supplier: Matrix Scientific

026697-500MG
101822-676EA 306.15 USD
101822-676
3-[4-(3-Chlorophenyl)piperazin-1-yl]propionic acid dihydrochloride
3-[4-(3-Chlorophenyl)piperazin-1-yl]propionic acid dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₃N₂O₂
MDL Number: MFCD09997694
CAS Number: 1185294-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR