5-Ethyl-4-thiophen-2-ylthiazol-2-ylamine dihydrochloride

Supplier: Matrix Scientific

026696-500MG
101822-674EA 306.15 USD
101822-674
5-Ethyl-4-thiophen-2-ylthiazol-2-ylamine dihydrochloride
5-Ethyl-4-thiophen-2-ylthiazol-2-ylamine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₂Cl₂N₂S₂
MW: 283.24 g/mol
MDL Number: MFCD11100516
CAS Number: 1185304-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR