4-Acetylamino-2,5-dimethylbenzenesulfonyl chloride

Supplier: Matrix Scientific

026689-500MG
101822-664EA 538.46 USD
101822-664
4-Acetylamino-2,5-dimethylbenzenesulfonyl chloride
4-Acetylamino-2,5-dimethylbenzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₃S
MW: 261.73 g/mol
MDL Number: MFCD08445689
CAS Number: 13632-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR