(4-Pyrrolidin-1-ylmethylphenoxy)acetic acid hydrochloride

Supplier: Matrix Scientific

026688-500MG
101822-662EA 306.15 USD
101822-662
(4-Pyrrolidin-1-ylmethylphenoxy)acetic acid hydrochloride
(4-Pyrrolidin-1-ylmethylphenoxy)acetic acid hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD09997689
CAS Number: 1185304-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR