4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid hydrochloride

Supplier: Matrix Scientific

026684-500MG
101822-654EA 495.38 USD
101822-654
4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid hydrochloride
4-[4-(2-Hydroxyethyl)piperazin-1-yl]benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD09997685
CAS Number: 1185000-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR