3-(2-Aminothiazol-4-yl)phenol hydrochloride

Supplier: Matrix Scientific

026682-500MG
101822-650EA 170.77 USD
101822-650
3-(2-Aminothiazol-4-yl)phenol hydrochloride
3-(2-Aminothiazol-4-yl)phenol hydrochloride

Hazard: Irritant

Formula: C₉H₉ClN₂OS
MDL Number: MFCD08582775
CAS Number: 1050556-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR