2-Chloromethyl-1H-benzoimidazole-5-carboxylic acid hydrochloride

Supplier: Matrix Scientific

026659-500MG
101822-628EA 252.31 USD
101822-628
2-Chloromethyl-1H-benzoimidazole-5-carboxylic acid hydrochloride
2-Chloromethyl-1H-benzoimidazole-5-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₉H₈Cl₂N₂O₂
MDL Number: MFCD07366485
CAS Number: 351226-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR