3-(2-Isopropylimidazol-1-yl)propionic acid hydrochloride

Supplier: Matrix Scientific

026653-500MG
101822-624EA 306.15 USD
101822-624
3-(2-Isopropylimidazol-1-yl)propionic acid hydrochloride
3-(2-Isopropylimidazol-1-yl)propionic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₅ClN₂O₂
MDL Number: MFCD09997682
CAS Number: 1052526-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR