4-Oxo-4-[N'-(thiophene-2-carbonyl)hydrazino]butyric acid

Supplier: Matrix Scientific

026637-500MG
101822-606EA 306.15 USD
101822-606
4-Oxo-4-[N'-(thiophene-2-carbonyl)hydrazino]butyric acid
4-Oxo-4-[N'-(thiophene-2-carbonyl)hydrazino]butyric acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₄S
MW: 242.26 g/mol
MDL Number: MFCD01961871
CAS Number: 314765-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR