(2-Methoxyethyl)(1-methylpiperidin-4-yl)amine dihydrochloride

Supplier: Matrix Scientific

026619-500MG
101822-586EA 306.15 USD
101822-586
(2-Methoxyethyl)(1-methylpiperidin-4-yl)amine dihydrochloride
(2-Methoxyethyl)(1-methylpiperidin-4-yl)amine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD09997677
CAS Number: 1185294-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR