2-Piperazin-1-ylmethyl-3H-quinazolin-4-one dihydrochloride

Supplier: Matrix Scientific

026598-500MG
101822-548EA 306.15 USD
101822-548
2-Piperazin-1-ylmethyl-3H-quinazolin-4-one dihydrochloride
2-Piperazin-1-ylmethyl-3H-quinazolin-4-one dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₄O
MDL Number: MFCD08166272
CAS Number: 725690-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR