5-Methyl-thiophene-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-Methylthiophene-3-carboxylic acid

026578-500MG
101822-512EA 116.92 USD
101822-512
5-Methyl-thiophene-3-carboxylic acid
5-Methyl-thiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₆O₂S
MW: 142.18 g/mol
MDL Number: MFCD01680229
CAS Number: 19156-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR