5-Ethyl-4-propylthiazol-2-ylamine hydrochloride

Supplier: Matrix Scientific

026575-500MG
101822-508EA 306.15 USD
101822-508
5-Ethyl-4-propylthiazol-2-ylamine hydrochloride
5-Ethyl-4-propylthiazol-2-ylamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₅ClN₂S
MW: 206.74 g/mol
MDL Number: MFCD09997658
CAS Number: 30709-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR