5-Methoxy-1-propyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

026568-500MG
101822-494EA 306.15 USD
101822-494
5-Methoxy-1-propyl-1H-indole-3-carbaldehyde
5-Methoxy-1-propyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD07186534
CAS Number: 128600-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR