2-Pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4,5-Dihydro-2-(pyridin-3-yl)-1,3-thiazole-4-carboxylic acid

026560-500MG
101822-482EA 306.15 USD
101822-482
2-Pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid
2-Pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂S
MW: 208.24 g/mol
MDL Number: MFCD02854711
CAS Number: 116247-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR