(2-Methoxyphenyl)hydrazine hydrochloride

Synonyms: 2-Methoxyphenylhydrazine hydrochloride2-Hydrazinoanisole hydrochloride

026556-500MG
101822-474EA 215.47 USD
101822-474
(2-Methoxyphenyl)hydrazine hydrochloride
(2-Methoxyphenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O
MW: 174.6280 g/mol
MDL Number: MFCD00035456
CAS Number: 6971-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR