4-Amino-1-isopropylpiperidine dihydrochloride

Supplier: Matrix Scientific

026555-500MG
101822-472EA 306.15 USD
101822-472
4-Amino-1-isopropylpiperidine dihydrochloride
4-Amino-1-isopropylpiperidine dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀N₂Cl₂
MDL Number: MFCD06796401
CAS Number: 534596-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR