3-(2,6-Dimethylmorpholin-4-yl)propionic acid hydrochloride

Supplier: Matrix Scientific

026551-500MG
101822-464EA 338.46 USD
101822-464
3-(2,6-Dimethylmorpholin-4-yl)propionic acid hydrochloride
3-(2,6-Dimethylmorpholin-4-yl)propionic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₃
MDL Number: MFCD09997648
CAS Number: 1185294-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR