3-(3-Methylpiperidin-1-yl)propionic acid hydrochloride

Supplier: Matrix Scientific

026550-500MG
101822-462EA 306.15 USD
101822-462
3-(3-Methylpiperidin-1-yl)propionic acid hydrochloride
3-(3-Methylpiperidin-1-yl)propionic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD09997647
CAS Number: 858444-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR