2-(Furan-2-ylmethoxy)phenylamine hydrochloride

Supplier: Matrix Scientific

026537-500MG
101822-442EA 306.15 USD
101822-442
2-(Furan-2-ylmethoxy)phenylamine hydrochloride
2-(Furan-2-ylmethoxy)phenylamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₂
MDL Number: MFCD09997638
CAS Number: 1185293-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR