[1,2,4]Triazolo[1,5-a]pyrimidin-2-ylamine hydrochloride

Supplier: Matrix Scientific

026534-500MG
101822-438EA 306.15 USD
101822-438
[1,2,4]Triazolo[1,5-a]pyrimidin-2-ylamine hydrochloride
[1,2,4]Triazolo[1,5-a]pyrimidin-2-ylamine hydrochloride

Hazard: Irritant

Formula: C₅H₆ClN₅
MDL Number: MFCD09997636
CAS Number: 1185294-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR