3-Dimethylaminomethyl-4-methoxybenzaldehyde hydrochloride

Supplier: Matrix Scientific

026509-500MG
101822-394EA 306.15 USD
101822-394
3-Dimethylaminomethyl-4-methoxybenzaldehyde hydrochloride
3-Dimethylaminomethyl-4-methoxybenzaldehyde hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₂
MDL Number: MFCD07357556
CAS Number: 1177308-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR