3-(2-Methylimidazol-1-ylmethyl)benzoic acid hydrochloride

Supplier: Matrix Scientific

026507-500MG
101822-390EA 181.54 USD
101822-390
3-(2-Methylimidazol-1-ylmethyl)benzoic acid hydrochloride
3-(2-Methylimidazol-1-ylmethyl)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₂
MDL Number: MFCD09997628
CAS Number: 1185293-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR