3-Imidazol-1-ylmethylbenzoic acid hydrochloride

Supplier: Matrix Scientific

026506-500MG
101822-388EA 495.38 USD
101822-388
3-Imidazol-1-ylmethylbenzoic acid hydrochloride
3-Imidazol-1-ylmethylbenzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂O₂
MDL Number: MFCD08690248
CAS Number: 218131-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR