(2-Chloro-6-fluorobenzyl)hydrazine hydrochloride

Supplier: Matrix Scientific

026497-500MG
101822-370EA 306.15 USD
101822-370
(2-Chloro-6-fluorobenzyl)hydrazine hydrochloride
(2-Chloro-6-fluorobenzyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₇H₉Cl₂FN₂
MDL Number: MFCD08458140
CAS Number: 1059626-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR