6,7,8,9-Tetrahydro-dibenzofuran-2-ylamine hydrochloride

Supplier: Matrix Scientific

026493-500MG
101822-362EA 306.15 USD
101822-362
6,7,8,9-Tetrahydro-dibenzofuran-2-ylamine hydrochloride
6,7,8,9-Tetrahydro-dibenzofuran-2-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄ClNO
MDL Number: MFCD03196914
CAS Number: 51595-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR