3-(3,5-Dimethylpyrazol-1-yl)benzaldehyde hydrochloride

Supplier: Matrix Scientific

026481-500MG
101822-342EA 306.15 USD
101822-342
3-(3,5-Dimethylpyrazol-1-yl)benzaldehyde hydrochloride
3-(3,5-Dimethylpyrazol-1-yl)benzaldehyde hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O
MDL Number: MFCD09997613
CAS Number: 1185293-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR