2-Ethyl-3-methyl-1H-indole-7-carboxylic acid

Supplier: Matrix Scientific

026468-500MG
101822-322EA 495.38 USD
101822-322
2-Ethyl-3-methyl-1H-indole-7-carboxylic acid
2-Ethyl-3-methyl-1H-indole-7-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD06755852
CAS Number: 876715-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR