3-(5-Fluoro-1H-benzoimidazol-2-yl)propylamine dihydrochloride

Supplier: Matrix Scientific

026466-500MG
101822-318EA 306.15 USD
101822-318
3-(5-Fluoro-1H-benzoimidazol-2-yl)propylamine dihydrochloride
3-(5-Fluoro-1H-benzoimidazol-2-yl)propylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄Cl₂FN₃
MDL Number: MFCD09029276
CAS Number: 883541-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR