4-Dimethylaminomethylbenzoic acid hydrochloride

Supplier: Matrix Scientific

Synonyms: 4-((Dimethylamino)methyl)benzoic acid hydrochloride

026462-500MG
101822-312EA 230.77 USD
101822-312
4-Dimethylaminomethylbenzoic acid hydrochloride
4-Dimethylaminomethylbenzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
MDL Number: MFCD09997604
CAS Number: 17847-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR