2-Morpholin-4-yl-2-phenylethylamine

Supplier: Matrix Scientific

026454-500MG
101822-298EA 170.77 USD
101822-298
2-Morpholin-4-yl-2-phenylethylamine
2-Morpholin-4-yl-2-phenylethylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
MDL Number: MFCD01631944
CAS Number: 31466-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR