2-Methylbenzofuran-5-ylamine hydrochloride

Supplier: Matrix Scientific

026446-500MG
101822-282EA 306.15 USD
101822-282
2-Methylbenzofuran-5-ylamine hydrochloride
2-Methylbenzofuran-5-ylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClNO
MDL Number: MFCD00035194
CAS Number: 23968-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR