Amino(3,5-dimethyl-1H-pyrazol-4-yl)acetic acid dihydrochloride

Supplier: Matrix Scientific

026439-500MG
101822-270EA 181.54 USD
101822-270
Amino(3,5-dimethyl-1H-pyrazol-4-yl)acetic acid dihydrochloride
Amino(3,5-dimethyl-1H-pyrazol-4-yl)acetic acid dihydrochloride

Hazard: Irritant

Formula: C₇H₁₃Cl₂N₃O₂
MW: 242.11 g/mol
MDL Number: MFCD09997591
CAS Number: 154108-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR