Adamantan-1-ylmethylmethylamine hydrobromide

Supplier: Matrix Scientific

026436-500MG
101822-264EA 306.15 USD
101822-264
Adamantan-1-ylmethylmethylamine hydrobromide
Adamantan-1-ylmethylmethylamine hydrobromide

Hazard: Irritant

Formula: C₁₂H₂₂BrN
MW: 260.22 g/mol
MDL Number: MFCD09997589
CAS Number: 1185301-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR